Trace Metals III for AA and ICP-OES
Lieferant: Honeywell Chemicals
E2168-100ML
HONCE2168-100MLEA
391.31
EUR
HONCE2168-100ML
Trace Metals III for AA and ICP-OES
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
500 mg/L each of Ba, Ca, Mo, Na, 100 mg/L each of K, Mg in HNO₃ 2%