internal standard mix for ICP
Lieferant: Honeywell Chemicals
E2172-100ML
HONCE2172-100MLEA 211.55 EUR
HONCE2172-100ML
internal standard mix for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
10 mg/L each of 6Li, Sc, Ge, Y, In, Tb, Bi in HNO₃ 5%