ELAN 6100 DRC setup stab masscal solution Hg for ICP
Lieferant: Honeywell Chemicals
E2182-100ML
HONCE2182-100MLEA 291.75 EUR
HONCE2182-100ML
ELAN 6100 DRC setup stab masscal solution Hg for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
10 g/L Ba; 1 g/L each of Al, Cd, Ce, Cr, Cu, In, Pb, Mg, Mn, Rh, Th in HNO₃ 0.5%