NexION setup solution
Lieferant: Honeywell Chemicals
E2188-100ML
HONCE2188-100MLEA
211.55
EUR
HONCE2188-100ML
NexION setup solution
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
1 g/L each of Be, Ce, Fe, In, Li, Mg, Pb, U in HNO₃ 1%