NexION KED mode detection limit solution
Lieferant: Honeywell Chemicals
E2194-100ML
HONCE2194-100MLEA 326.32 EUR
HONCE2194-100ML
NexION KED mode detection limit solution
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
10 g/L each of V, As, Se in HCl 1%