NexION AFT single-element solution
Lieferant: Honeywell Chemicals
E2196-100ML
HONCE2196-100MLEA 120.29 EUR
HONCE2196-100ML
NexION AFT single-element solution
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
2 g/L Iron (Fe) in HNO₃ 2%