NexION AFT multi-element solution
Lieferant: Honeywell Chemicals
E2197-100ML
HONCE2197-100MLEA 1078.5 EUR
HONCE2197-100ML
NexION AFT multi-element solution
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
2 g/L each of Ag, Al, As, Ba, Be, Bi, Ca, Cd, Cr, Co, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn in HNO₃ 2%