NexION KED mode detection limit solution
Lieferant: Honeywell Chemicals
E2198-100ML
HONCE2198-100MLEA 152.09 EUR
HONCE2198-100ML
NexION KED mode detection limit solution
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
Blank Solution in HCl 1%