ICP standard solutions, Fluka™
Lieferant: Honeywell Chemicals
E2213-100ML E2214-100ML E2216-100ML E2217-100ML E2218-100ML
HONCE2213-100MLEA 482.57 EUR
HONCE2213-100ML HONCE2214-100ML HONCE2216-100ML HONCE2217-100ML HONCE2218-100ML
ICP standard solutions, Fluka™
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Materials (CRMs).
  • Traceable to NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)