Multi-element standard solution 3 for ICP
Lieferant: Honeywell Chemicals
E2231-100ML
HONCE2231-100MLEA
116.28
EUR
HONCE2231-100ML
Multi-element standard solution 3 for ICP
Standards
Kationischer Standards
Mehrelement-Standards
Certified Reference Material (CRM).
2000 mg/L Ca, 1000 mg/L Na, 400 mg/L Mg, 200 mg/L K in HNO₃ 5%