Ammonium Standard for Ion Chromatography, (CRM), Fluka™
Lieferant: Honeywell Chemicals
E3001-100ML
HONCE3001-100MLEA
0
EUR
HONCE3001-100ML
Ammonium Standard for Ion Chromatography, (CRM), Fluka™
Standards
Kationischer Standards
Ammonium-Standards
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).