Bromate standard for ion chromatography, (CRM), Fluka™
Lieferant: Honeywell Chemicals
E3005-100ML
HONCE3005-100MLEA 54.62 EUR
HONCE3005-100ML
Bromate standard for ion chromatography, (CRM), Fluka™
Standards Anionische Standards Bromat-Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).