Lithium standard for ion chromatography, (CRM), Fluka™
Lieferant: Honeywell Chemicals
E3024-100ML
HONCE3024-100MLEA 54.62 EUR
HONCE3024-100ML
Lithium standard for ion chromatography, (CRM), Fluka™
Standards Kationischer Standards Lithium-Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).