Nitrate standard for ion chromatography, (CRM), Fluka™
Lieferant: Honeywell Chemicals
E3031-100ML
HONCE3031-100MLEA
54.62
EUR
HONCE3031-100ML
Nitrate standard for ion chromatography, (CRM), Fluka™
Standards
Anionische Standards
Nitrat-Standards
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).