Phosphate (P) Standard for Ion chromatography, (CRM), Fluka™
Lieferant: Honeywell Chemicals
E3040-100ML
HONCE3040-100MLEA
0
EUR
HONCE3040-100ML
Phosphate (P) Standard for Ion chromatography, (CRM), Fluka™
Standards
Anionische Standards
Phosphat-Standards
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).