Tartrate Standard for Ion Chromatography, (CRM), Fluka™
Lieferant: Honeywell Chemicals
E3049-100ML
HONCE3049-100MLEA 52.12 EUR
HONCE3049-100ML
Tartrate Standard for Ion Chromatography, (CRM), Fluka™
Standards Anionische Standards Tartrat-Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).