Triethanolamine Standard for Ion Chromatography, (CRM), Fluka™
Lieferant: Honeywell Chemicals
E3052-100ML
HONCE3052-100MLEA 85.17 EUR
HONCE3052-100ML
Triethanolamine Standard for Ion Chromatography, (CRM), Fluka™
Standards Kationischer Standards Amin-Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).