Cadmium AAS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4014-100ML E4014-500ML E4015-100ML E4015-500ML
HONCE4014-100MLEA 34.01 EUR
HONCE4014-100ML HONCE4014-500ML HONCE4015-100ML HONCE4015-500ML
Cadmium AAS standards, (CRMs), Fluka™
Standards Kationischer Standards Cadmium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).