Cadmium AAS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4014-100ML
E4014-500ML
E4015-100ML
E4015-500ML
HONCE4014-100MLEA
34.01
EUR
HONCE4014-100ML
HONCE4014-500ML
HONCE4015-100ML
HONCE4015-500ML
Cadmium AAS standards, (CRMs), Fluka™
Standards
Kationischer Standards
Cadmium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).