Europium AAS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4026-100ML
E4026-500ML
HONCE4026-100MLEA
85.73
EUR
HONCE4026-100ML
HONCE4026-500ML
Europium AAS Standards, (CRMs), Fluka™
Standards
Kationischer Standards
Europium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).