Gadolinium AAS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4030-500ML
HONCE4030-500MLEA
120.29
EUR
HONCE4030-500ML
Gadolinium AAS Standards, (CRMs), Fluka™
Standards
Kationischer Standards
Gadolinium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).