Iridium AAS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4036-500ML
HONCE4036-500MLEA 721.78 EUR
HONCE4036-500ML
Iridium AAS Standards, (CRMs), Fluka™
Standards Kationischer Standards Iridium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).