Magnesium AAS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4042-100ML
E4042-500ML
E4043-100ML
E4043-500ML
HONCE4042-100MLEA
30.02
EUR
HONCE4042-100ML
HONCE4042-500ML
HONCE4043-100ML
HONCE4043-500ML
Magnesium AAS standards, (CRMs), Fluka™
Standards
Kationischer Standards
Magnesium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).