Nickel AAS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4053-500ML
HONCE4053-500MLEA
56
EUR
HONCE4053-500ML
Nickel AAS standards, (CRMs), Fluka™
Standards
Kationischer Standards
Nickel-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).