Nickel AAS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4053-500ML
HONCE4053-500MLEA 56 EUR
HONCE4053-500ML
Nickel AAS standards, (CRMs), Fluka™
Standards Kationischer Standards Nickel-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).