Rubidium AAS Standards, (CRMs), Fluka™
E4059-500ML
HONCE4059-500MLEA 89.32 EUR
HONCE4059-500ML
Rubidium AAS Standards, (CRMs), Fluka™
Standards Kationischer Standards Rubidium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).