Samarium AAS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4069-500ML
HONCE4069-500MLEA 107.85 EUR
HONCE4069-500ML
Samarium AAS Standards, (CRMs), Fluka™
Standards Kationischer Standards Samarium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).