Calcium AAS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4012-100ML E4012-500ML E4013-100ML E4013-500ML E4107-100ML E4108-100ML E4200-100ML
HONCE4012-100MLEA 34.99 EUR
HONCE4012-100ML HONCE4012-500ML HONCE4013-100ML HONCE4013-500ML HONCE4107-100ML HONCE4108-100ML HONCE4200-100ML
Calcium AAS standards, (CRMs), Fluka™
Standards Kationischer Standards Calcium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).