Calcium AAS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4012-100ML
E4012-500ML
E4013-100ML
E4013-500ML
E4107-100ML
E4108-100ML
E4200-100ML
HONCE4012-100MLEA
34.99
EUR
HONCE4012-100ML
HONCE4012-500ML
HONCE4013-100ML
HONCE4013-500ML
HONCE4107-100ML
HONCE4108-100ML
HONCE4200-100ML
Calcium AAS standards, (CRMs), Fluka™
Standards
Kationischer Standards
Calcium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).