Silicon AAS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4119-100ML
HONCE4119-100MLEA
80.2
EUR
HONCE4119-100ML
Silicon AAS Standards, (CRMs), Fluka™
Standards
Kationischer Standards
Silizium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).