Silicon AAS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4119-100ML
HONCE4119-100MLEA 80.2 EUR
HONCE4119-100ML
Silicon AAS Standards, (CRMs), Fluka™
Standards Kationischer Standards Silizium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).