Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Lieferant: Honeywell Chemicals
E4300-100ML E4300-500ML E4301-100ML E4302-100ML
HONCE4300-100MLEA 248.33 EUR
HONCE4300-100ML HONCE4300-500ML HONCE4301-100ML HONCE4302-100ML
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Puffer Ionisationspuffer
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)
Lieferumfang: Supplied complete with a detailed Certificate of Analysis (CoA) in every delivery.