Analytical standard, Simeton, reference material, Honeywell Fluka™
Lieferant: Honeywell Chemicals
R1173-25MG
HONCR1173-25MGEA
164.99
EUR
HONCR1173-25MG
Analytical standard, Simeton, reference material, Honeywell Fluka™
Standards
Organische Standards
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results