Analytical standard, Desoxycarbadox-(methyl-d₃), reference material, Honeywell Fluka™
Lieferant: Honeywell Chemicals
R1208-10MG
HONCR1208-10MGEA 322.63 EUR
HONCR1208-10MG
Analytical standard, Desoxycarbadox-(methyl-d₃), reference material, Honeywell Fluka™
Standards Organische Standards
  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results