Analytical standard, Flumequine-(1,2,carboxy-13C3), reference material, Honeywell Fluka™
Lieferant: Honeywell Chemicals
R1233-10MG
HONCR1233-10MGEA 941.62 EUR
HONCR1233-10MG
Analytical standard, Flumequine-(1,2,carboxy-13C3), reference material, Honeywell Fluka™
Standards Organische Standards
  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results