ASTM® D6584 1,2,4-Butanetriol solution, 1000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
Lieferant: Honeywell Chemicals
R1384-5ML
HONCR1384-5MLEA 356.2 EUR
HONCR1384-5ML
ASTM® D6584 1,2,4-Butanetriol solution, 1000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
Standards Mineralölindustrie-Standards
High quality inorganic and organic reference materials produced according to ISO 9001
  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results