ASTM® D6584 Tricaprin solution, 8000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
Lieferant: Honeywell Chemicals
R1385-5ML
HONCR1385-5MLEA 233.57 EUR
HONCR1385-5ML
ASTM® D6584 Tricaprin solution, 8000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
Standards Mineralölindustrie-Standards
High quality inorganic and organic reference materials produced according to ISO 9001.
  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results