ASTM® D6584 individual stock and internal standards mix Kit, analytical standard, Honeywell Fluka™
Lieferant: Honeywell Chemicals
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
Kit contains:
R1380-1ML ASTM® D6584 Glycerin solution, 500 μg/ml in pyridine, reference material
R1381-3ML ASTM® D6584 Monoolein solution, 5000 μg/ml in pyridine, reference material
R1382-2ML ASTM® D6584 1,3-Diolein solution, 5000 μg/ml in pyridine, reference material
R1383-2ML ASTM® D6584 Triolein solution, 5000 μg/ml in pyridine, reference material
R1384-5ML ASTM® D6584 Butanetriol solution, 1000 μg/ml in pyridine, reference material
R1385-5ML ASTM® D6584 Tricaprin solution, 8000 μg/ml in pyridine, reference material