Aflatoxin B2-solution 0.5 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
Lieferant: Honeywell Chemicals
R1957-2ML
HONCR1957-2MLEA 260.06 EUR
HONCR1957-2ML
Aflatoxin B2-solution 0.5 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
Standards Organische Standards
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery