Kaliumhydrogendifluorid
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Kaliumhydrogenfluorid, Kaliumbifluorid
01138.9025
HONE01138.9025EA
0
EUR
HONE01138.9025
Kaliumhydrogendifluorid
Potassium hydrogen difluoride
Formel:
HF₂K Molecular Weight: 78,1 g/mol Schmelzpunkt: 239 °C Dichte: 2,37 g/cm³ (20 °C) |
MDL:
MFCD00011403 CAS-Nummer: 7789-29-9 EINECS: 232-156-2 UN: 1811 ADR: 8,II Merck Index: 12,07771 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...