Nickel(II)fluorid Tetrahydrat

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Nickeldifluorid Tetrahydrat

01258.9050
HONE01258.9050EA 0 EUR
HONE01258.9050
Nickel(II)fluorid Tetrahydrat
Nickel(II)fluorid Tetrahydrat
Formel: F₂Ni·4H₂O
Molecular Weight: 168,75 g/mol
MDL: MFCD00016263
CAS-Nummer: 13940-83-5
EINECS: 233-071-3
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR