Aluminiumfluorid
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Aluminum fluoride
01277.9040
HONE01277.9040EA
0
EUR
HONE01277.9040
Aluminiumfluorid
Aluminium fluoride
Formel:
AlF₃ Schmelzpunkt: 1040 °C Dichte: 2,88 g/cm³ (20 °C) |
MDL:
MFCD00003426 CAS-Nummer: 7784-18-1 EINECS: 232-051-1 UN: 3260 ADR: 8,II Merck Index: 12,00351 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...