Ammoniumhexafluorosilicat

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Ammoniumfluorosilicat

01401.9025
HONE01401.9025EA 0 EUR
HONE01401.9025
Ammoniumhexafluorosilicat
Ammoniumhexafluorosilicat
Formel: H₈F₆N₂Si
Molecular Weight: 178,15 g/mol
Dichte: 2,01 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010887
CAS-Nummer: 16919-19-0
EINECS: 240-968-3
UN: 2854
ADR: 6.1,III
Merck Index: 13,00529

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR