Magnesiumhexafluorosilicat Hexahydrat

Lieferant: Honeywell Chemicals

01415.9025
HONE01415.9025EA 0 EUR
HONE01415.9025
Magnesiumhexafluorosilicat Hexahydrat
Magnesiumhexafluorosilicat Hexahydrat
Formel: F₆MgSi·6H₂O
MDL: MFCD00150250
CAS-Nummer: 18972-56-0
EINECS: 241-022-2
UN: 2853
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR