Natriumhexafluorosilicat
Lieferant: Honeywell Chemicals
|
Gefahr
|
01418.9050
HONE01418.9050EA
0
EUR
HONE01418.9050
Natriumhexafluorosilicat
Natriumhexafluorosilicat
Formel:
F₆Na₂Si Molecular Weight: 188,06 g/mol Dichte: 2,68 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003491 CAS-Nummer: 16893-85-9 EINECS: 240-934-8 UN: 2674 ADR: 6.1,III Merck Index: 12,08769 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...