Natriumtetrafluoroborat

Lieferant: Honeywell Chemicals

Synonyme: Sodium fluoroborate

01509.9050
HONE01509.9050EA 0 EUR
HONE01509.9050
Natriumtetrafluoroborat
Sodium tetrafluoroborate
Formel: BF₄Na
Molecular Weight: 109,79 g/mol
Schmelzpunkt: 384 °C
Dichte: ∼2,47 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003515
CAS-Nummer: 13755-29-8
EINECS: 237-340-6
Merck Index: 12,08761

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR