Kaliumhexafluorozirkonat

Lieferant: Honeywell Chemicals

01712.9050
HONE01712.9050EA 0 EUR
HONE01712.9050
Kaliumhexafluorozirkonat
Potassium hexafluorozirconate
Formel: F₆K₂Zr
MDL: MFCD00042547
CAS-Nummer: 16923-95-8
EINECS: 240-985-6
UN: 3288
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR