4,4'-(Hexafluorisopropyliden)diphenol

Lieferant: Honeywell Chemicals

Synonyme: 4,4′-(Hexafluoroisopropylidene)diphenol, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, 4,4'-(Perfluoropropane-2,2-diyl)diphenol

01821.9010
HONE01821.9010EA 0 EUR
HONE01821.9010
4,4'-(Hexafluorisopropyliden)diphenol
4,4'-(Hexafluorisopropyliden)diphenol
Formel: C₁₅H₁₀F₆O₂
Molecular Weight: 336,24 g/mol
MDL: MFCD00000439
CAS-Nummer: 1478-61-1
EINECS: 216-036-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR