Sodium hexametaphosphate
Lieferant: Honeywell Chemicals
Synonyme:
Polyphosphate sodium salt, Sodium polyphosphate, Calgon, Phosphate glass, water soluble, SHMP
04267.9025
HONE04267.9025EA
0
EUR
HONE04267.9025
Sodium hexametaphosphate
Sodium hexametaphosphate
Formel:
(NaO₃P)n Dichte: 2,4 g/cm³ (20 °C) |
MDL:
MFCD00134118 CAS-Nummer: 68915-31-1 EINECS: 256-779-4 Merck Index: 12,08814 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...