Kaliumphosphinat

Lieferant: Honeywell Chemicals

04411.9050
HONE04411.9050EA 0 EUR
HONE04411.9050
Kaliumphosphinat
Potassium phosphinate
Formel: H₂KO₂P
CAS-Nummer: 7782-87-8
EINECS: 231-967-9
Merck Index: 12,07807

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR