Yttrium(III)fluorid
Lieferant: Honeywell Chemicals
Synonyme:
Yttriumtrifluorid
10417.9025
HONE10417.9025EA
0
EUR
HONE10417.9025
Yttrium(III)fluorid
Yttrium(III)fluorid
Formel:
F₃Y Molecular Weight: 145,9 g/mol Siedepunkt: 2227 °C (1013 hPa) Schmelzpunkt: 1387 °C Dichte: 4,01 g/cm³ (20 °C) |
MDL:
MFCD00049714 CAS-Nummer: 13709-49-4 EINECS: 237-257-5 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...