Aluminiumchlorid Hexahydrat
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Aluminum chloride hexahydrate
11016.9025
HONE11016.9025EA
0
EUR
HONE11016.9025
Aluminiumchlorid Hexahydrat
Aluminiumchlorid Hexahydrat
Formel:
AlCl₃·6H₂O Molecular Weight: 241,43 g/mol Schmelzpunkt: ∼100 °C Dichte: 2,39 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149134 CAS-Nummer: 7784-13-6 EINECS: 231-208-1 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...