Aluminiumoxid ≥98% (Al₂O₃-Basis), puriss., reinst, Fluka™
Lieferant: Honeywell Chemicals
11028.9025
HONE11028.9025EA
0
EUR
HONE11028.9025
Aluminiumoxid ≥98% (Al₂O₃-Basis), puriss., reinst, Fluka™
Aluminium oxide
Formel:
Al₂O₃ Siedepunkt: 2980 °C (1013 hPa) Schmelzpunkt: 2045 °C Dichte: 3,94 g/cm³ (20 °C) |
MDL:
MFCD00003424 CAS-Nummer: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 13,00355 |
Spezifikation Testergebnisse
Assay of Al2O₃ | Min. 98 % |
Loss on ignition (1000°C, 2 h) | Max. 0.5 % |
Alkalis (as Na₂O) | Max. 0.2 % |
Iron (Fe) | Max. 0.03 % |
Heavy metals (as Pb) | Max. 0.005 % |
Chloride (Cl) | Max. 0.005 % |
Silicate (as SiO₂) | Max. 0.1 % |
Sulfate (SO₄) | Max. 0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...