Ammoniumsulfat

Lieferant: Honeywell Chemicals

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

11225.9025
HONE11225.9025EA 0 EUR
HONE11225.9025
Ammoniumsulfat
Ammoniumsulfat
Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
Schmelzpunkt: 336…339 °C
Dichte: 1.78 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR